siRNA Details
Gene Symbol: Tet2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233347.2 | XP_006233409.1 | 11 | 4709 | ||
XM_008761534.1 | XP_008759756.1 | 11 | 4706 | |||
XM_227694.7 | XP_227694.5 | 10 | 4592 |
