siRNA Details
Gene Symbol: Alpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761482.1 | XP_008759704.1 | 11 | 2256 | ||
XM_008761483.1 | XP_008759705.1 | 10 | 1836 | |||
XM_227715.7 | XP_227715.5 | 10 | 2363 |
