siRNA Details
Gene Symbol: Alpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761482.1 | XP_008759704.1 | 12 | 3770 | ||
XM_008761483.1 | XP_008759705.1 | 11 | 3350 | |||
XM_227715.7 | XP_227715.5 | 11 | 3877 |

More Information
Other Species Targeted

Gene Ontology Categories:
