siRNA Details
Gene Symbol: Gpr107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107828.1 | NP_001101298.1 | 4 | 344 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_178760, AK142454, AK034815, BC092231, AK083025, and others.
Set Membership: |
![]() |
