siRNA Details
Gene Symbol: Cln6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191794.1 | NP_001178723.1 | 2 | 158 | ||
XM_006243238.2 | XP_006243300.1 | 3 | 248 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001033175, AK154551, BC116794, AK154414, BC144722, and others.
Gene Ontology Categories:
