siRNA Details
Gene Symbol: Ankrd23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108211.1 | NP_001101681.1 | 3, 4 | 469 | ||
XM_006244814.2 | XP_006244876.1 | 2, 3 | 232 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ankrd23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108211.1 | NP_001101681.1 | 3, 4 | 469 | ||
XM_006244814.2 | XP_006244876.1 | 2, 3 | 232 |