siRNA Details
Gene Symbol: Pdcl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025709.1 | NP_001020880.1 | 5 | 549 | ||
GenBank mRNA | BC099162.1 | - | 5 | 549 | 7381935 | |
FQ211568.1 | - | 658 | ||||
FQ229403.1 | - | 5 | 534 |
