siRNA Details
Gene Symbol: Pdcl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025709.1 | NP_001020880.1 | 6 | 676 | ||
GenBank mRNA | BC099162.1 | - | 6 | 676 | 7381935 | |
FQ211568.1 | - | 785 | ||||
FQ229403.1 | - | 6 | 661 |
