siRNA Details
Gene Symbol: Rab40c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182675.1 | NP_872616.1 | 1 | 101 | ||
XM_006245981.2 | XP_006246043.2 | 2 | 198 | |||
GenBank mRNA | AY190240.1 | - | 1 | 101 | ||
BC128720.1 | - | 1 | 142 | 8374477 |
