siRNA Details
Gene Symbol: Mrps23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108289.1 | NP_001101759.1 | 2 | 216 | ||
XM_008768075.1 | XP_008766297.1 | 2 | 407 | |||
GenBank mRNA | FQ213501.1 | - | 2 | 752 | ||
FQ216939.1 | - | 2 | 247 |
