siRNA Details
Gene Symbol: Rpo1-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037787.1 | NP_001032876.1 | 3 | 261 | ||
GenBank mRNA | BC098735.1 | - | 3 | 261 | 7385125 | |
FQ221941.1 | - | 3 | 338 | |||
FQ232634.1 | - | 3 | 320 |
