siRNA Details
Gene Symbol: Cpeb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770226.1 | XP_008768448.1 | 1545 | |||
GenBank mRNA | JF973322.1 | - | 1455 | |||
JF973323.1 | - | 1545 |

More Information
Other Species Targeted

Gene Ontology Categories:
