siRNA Details
Gene Symbol: Ccdc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108382.1 | NP_001101852.1 | 8 | 678 | ||
XM_006252132.2 | XP_006252194.1 | 8 | 677 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108382.1 | NP_001101852.1 | 8 | 678 | ||
XM_006252132.2 | XP_006252194.1 | 8 | 677 |