siRNA Details
Gene Symbol: Pbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108399.1 | NP_001101869.1 | 6 | 853 | ||
XM_006252909.2 | XP_006252971.1 | 7 | 844 | |||
XM_008771129.1 | XP_008769351.1 | 6 | 948 | |||
XM_008771130.1 | XP_008769352.1 | 6 | 706 |

More Information
Other Species Targeted

Gene Ontology Categories:
