siRNA Details
Gene Symbol: Ccny | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191833.1 | NP_001178762.1 | 6 | 421 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026484, AK076556, BC023321, AK028696, AK017493, and others.
Gene Ontology Categories:
