siRNA Details
Gene Symbol: Ginm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227626.2 | XP_006227688.1 | 7 | 922 | ||
XM_006227627.2 | XP_006227689.1 | 7 | 916 | |||
XM_341726.7 | XP_341727.4 | 7 | 920 | |||
GenBank mRNA | FQ228838.1 | - | 715 |
