siRNA Details
Gene Symbol: Mfsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191847.1 | NP_001178776.1 | 2, 3 | 238 | ||
XM_008761056.1 | XP_008759278.1 | 2, 3 | 240 | |||
GenBank mRNA | FQ211604.1 | - | 255 | |||
FQ217493.1 | - | 223 |
