siRNA Details
Gene Symbol: Trim44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013203.1 | NP_001013221.1 | 1 | 640 | ||
GenBank mRNA | AY551091.1 | - | 1 | 640 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trim44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013203.1 | NP_001013221.1 | 1 | 640 | ||
GenBank mRNA | AY551091.1 | - | 1 | 640 |