siRNA Details
Gene Symbol: Rsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014193.2 | NP_001014215.2 | 3 | 816 | ||
GenBank mRNA | BC085888.1 | - | 3 | 764 | 7192402 | |
CO405856.1 | - | 2 | 578 | |||
FQ230600.1 | - | 3 | 801 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
