siRNA Details
Gene Symbol: Nfam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242140.2 | XP_006242202.1 | 4 | 766 | ||
XM_006242142.2 | XP_006242204.1 | 4 | 679 |

More Information
Other Species Targeted

Gene Ontology Categories:
