siRNA Details
Gene Symbol: Arpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135045.1 | NP_001128517.1 | 5, 6 | 666 | ||
XM_006243972.2 | XP_006244034.1 | 6, 7 | 838 | |||
XM_006243973.2 | XP_006244035.1 | 6, 7 | 829 | |||
XM_006243974.2 | XP_006244036.1 | 6, 7 | 1254 | |||
XM_006243975.2 | XP_006244037.1 | 6, 7 | 901 | |||
XM_006243976.2 | XP_006244038.1 | 6, 7 | 787 | |||
XM_006243977.2 | XP_006244039.1 | 6, 7 | 997 | |||
XM_006243980.2 | XP_006244042.1 | 6, 7 | 867 | |||
XM_006243982.2 | XP_006244044.1 | 5, 6 | 941 | |||
XM_008766633.1 | XP_008764855.1 | 6, 7 | 4751 | |||
XM_008766634.1 | XP_008764856.1 | 7, 8 | 4600 | |||
XM_008766635.1 | XP_008764857.1 | 6, 7 | 4432 | |||
XM_008766636.1 | XP_008764858.1 | 6, 7 | 4751 | |||
XM_008766637.1 | XP_008764859.1 | 6, 7 | 4751 | |||
XM_008766638.1 | XP_008764860.1 | 6, 7 | 4751 | |||
XM_008766639.1 | XP_008764861.1 | 6, 7 | 4751 | |||
XM_008766640.1 | XP_008764862.1 | 6, 7 | 4751 | |||
XM_008766641.1 | XP_008764863.1 | 6, 7 | 4751 | |||
XM_008766642.1 | XP_008764864.1 | 6, 7 | 4751 | |||
XM_008766643.1 | XP_008764865.1 | 6, 7 | 4751 | |||
XM_008766644.1 | XP_008764866.1 | 6, 7 | 4751 |
