siRNA Details
Gene Symbol: Rhox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135607.1 | NP_001129079.1 | 760 | |||
XM_008773382.1 | XP_008771604.1 | 561 | ||||
XM_008773383.1 | XP_008771605.1 | 3, 4 | 507 | |||
XM_008773384.1 | XP_008771606.1 | 446 | ||||
GenBank mRNA | DQ058651.1 | - | 760 |
Gene Symbol: LOC103690001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257477.2 | XP_006257539.1 | 3, 4 | 507 | ||
XM_006257478.2 | XP_006257540.1 | 2, 3 | 446 | |||
XM_008773510.1 | XP_008771732.1 | 3, 4 | 561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
