siRNA Details
Gene Symbol: Map2k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025425.1 | NP_001020596.1 | 2, 3 | 274 | ||
XM_006248768.2 | XP_006248830.1 | 3, 4 | 459 | |||
XM_006248769.1 | XP_006248831.1 | 3, 4 | 464 | |||
XM_008768985.1 | XP_008767207.1 | 2, 3 | 415 | |||
GenBank mRNA | AY879265.1 | - | 2, 3 | 274 | ||
GU264001.1 | - | 3, 4 | 322 |
