siRNA Details
Gene Symbol: Smim14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037792.1 | NP_001032881.1 | 5 | 790 | ||
XM_006250961.2 | XP_006251023.1 | 5 | 1048 | |||
GenBank mRNA | BC100271.1 | - | 5 | 790 | 7445571 | |
FQ229716.1 | - | 5 | 850 | |||
FQ229847.1 | - | 4 | 675 |
