siRNA Details
Gene Symbol: Polr3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014259.1 | NP_001014281.1 | 3 | 819 | ||
GenBank mRNA | BC089990.1 | - | 3 | 819 | 7319381 | |
FQ212572.1 | - | 3 | 841 | |||
FQ213710.1 | - | 3 | 842 | |||
FQ213830.1 | - | 3 | 853 | |||
FQ216547.1 | - | 3 | 842 | |||
FQ226947.1 | - | 3 | 824 | |||
U53183.1 | - | 1 | 68 |
