siRNA Details
Gene Symbol: Rab9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109018.1 | NP_001102488.1 | 3 | 743 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_176971, AK049693, BC107383, AB232603, AK161867, and others.
Gene Ontology Categories:
