siRNA Details
Gene Symbol: Olr278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000730.1 | NP_001000730.1 | 1 | 447 |
Gene Symbol: Olr279 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000226.1 | NP_001000226.1 | 1 | 447 |
