siRNA Details
Gene Symbol: LOC100911042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751070.3 | XP_003751118.1 | 3 | 356 |
Gene Symbol: Smco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248472.2 | XP_006248534.1 | 358 | |||
GenBank mRNA | FQ217149.1 | - | 356 |
