siRNA Details
Gene Symbol: LOC102552640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770611.1 | XP_008768833.1 | 5 | 694 |
Gene Symbol: Rcor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013994.1 | NP_001014016.1 | 5 | 700 | ||
XM_008760113.1 | XP_008758335.1 | 6 | 550 | |||
XM_008760114.1 | XP_008758336.1 | 6 | 617 | |||
XM_008760115.1 | XP_008758337.1 | 6 | 736 | |||
GenBank mRNA | BC089209.1 | - | 5 | 700 | 7302520 |
