siRNA Details
Gene Symbol: Mapk1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122782.1 | NP_001116254.1 | 1 | 1225 | ||
XM_006230487.2 | XP_006230549.1 | 3 | 2369 | |||
XM_006230488.2 | XP_006230550.1 | 2 | 2660 | |||
XM_006230489.2 | XP_006230551.1 | 4 | 1534 | |||
XM_006230490.2 | XP_006230552.1 | 3 | 1411 | |||
XM_006230491.2 | XP_006230553.1 | 2 | 1290 | |||
XM_008760006.1 | XP_008758228.1 | 3 | 1825 | |||
XM_008760007.1 | XP_008758229.1 | 2 | 1702 | |||
GenBank mRNA | BC158819.1 | - | 1 | 1225 | 7374980 |
