siRNA Details
Gene Symbol: M1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236719.2 | XP_006236781.1 | 4 | 623 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_033079, BC119803, AK131954, BC122521, AF084364.
Gene Ontology Categories:
