siRNA Details
Gene Symbol: Mcmdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024340.1 | NP_001019511.2 | 2, 3 | 131 | ||
XM_006237781.2 | XP_006237843.1 | 1, 2 | 2957 | |||
XM_006237782.2 | XP_006237844.1 | 2, 3 | 760 | |||
XM_006237784.2 | XP_006237846.1 | 2, 3 | 2512 | |||
XM_006237785.2 | XP_006237847.1 | 1, 2 | 2959 | |||
XM_006237787.2 | XP_006237849.1 | 1, 2 | 2956 | |||
XM_006237788.2 | XP_006237850.1 | 1, 2 | 2954 | |||
XM_006237789.2 | XP_006237851.1 | 1, 2 | 2954 | |||
GenBank mRNA | BC083882.1 | - | 2, 3 | 131 | 7131459 |

More Information
Other Species Targeted

Gene Ontology Categories:
