siRNA Details
Gene Symbol: Mcmdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024340.1 | NP_001019511.2 | 15 | 1951 | ||
XM_006237781.2 | XP_006237843.1 | 14 | 4777 | |||
XM_006237782.2 | XP_006237844.1 | 15 | 2580 | |||
XM_006237784.2 | XP_006237846.1 | 15 | 4332 | |||
XM_006237785.2 | XP_006237847.1 | 14 | 4764 | |||
XM_006237788.2 | XP_006237850.1 | 12 | 4521 | |||
GenBank mRNA | BC083882.1 | - | 15 | 1951 | 7131459 |
