siRNA Details
Gene Symbol: Bbs10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109286.1 | NP_001102756.1 | 2 | 1618 | ||
XM_006241323.2 | XP_006241385.1 | 2 | 1584 | |||
GenBank mRNA | BC160915.1 | - | 2 | 1610 | 7457522 | |
FQ211675.1 | - | 2 | 1600 |
