siRNA Details
Gene Symbol: Cd84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192006.1 | NP_001178935.1 | 4, 5 | 792 | ||
XM_008769753.1 | XP_008767975.1 | 5, 6 | 1207 | |||
XM_008769754.1 | XP_008767976.1 | 6, 7 | 1274 | |||
XM_008769755.1 | XP_008767977.1 | 5, 6 | 1185 | |||
XM_008769757.1 | XP_008767979.1 | 4, 5 | 810 |

More Information
Other Species Targeted

Gene Ontology Categories:
