siRNA Details
Gene Symbol: Dnajc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134640.1 | NP_001128112.1 | 2, 3 | 188 | ||
XM_006232259.2 | XP_006232321.1 | 2, 3 | 203 | |||
GenBank mRNA | FQ212146.1 | - | 2, 3 | 147 |

More Information
Other Species Targeted

Gene Ontology Categories:
