siRNA Details
Gene Symbol: Srgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020074.2 | NP_064459.1 | 3 | 487 | ||
XM_006256483.2 | XP_006256545.1 | 6 | 867 | |||
XM_006256484.2 | XP_006256546.1 | 4 | 675 | |||
GenBank mRNA | BC088144.1 | - | 3 | 487 | 7384101 | |
FQ220127.1 | - | 3 | 506 | |||
FQ227535.1 | - | 3 | 504 | |||
FQ231972.1 | - | 3 | 504 | |||
J03224.1 | - | 3 | 500 | |||
K02934.1 | - | 3 | 502 |
