siRNA Details
Gene Symbol: Max | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022210.1 | NP_071546.1 | 2 | 48 | ||
XM_008764846.1 | XP_008763068.1 | 2 | 279 | |||
GenBank mRNA | BC099778.1 | - | 2 | 210 | 7933028 | |
D14447.1 | - | 2 | 48 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008558, S80521, BC138671, BC138672.
Gene Ontology Categories:
