siRNA Details
Gene Symbol: Fxyd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022388.1 | NP_071783.1 | 8 | 1292 | ||
XM_006237206.2 | XP_006237268.1 | 9 | 631 | |||
XM_008763260.1 | XP_008761482.1 | 8 | 1048 | |||
XM_008763261.1 | XP_008761483.1 | 8 | 1006 | |||
GenBank mRNA | L41254.1 | - | 8 | 1292 |
