siRNA Details
Gene Symbol: LOC100909604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762944.1 | XP_008761166.1 | 3 | 3133 | ||
XM_008762945.1 | XP_008761167.1 | 2 | 5382 |
Gene Symbol: Hoxa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236513.2 | XP_006236575.1 | 3 | 2510 |
Gene Symbol: LOC100911511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752179.3 | XP_003752227.1 | 2 | 3327 | ||
XM_006257703.2 | XP_006257765.1 | 3 | 3133 |
