siRNA Details
Gene Symbol: Hspb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131002.1 | NP_001124474.1 | 5 | 414 | ||
XM_006238524.2 | XP_006238586.2 | 5 | 420 | |||
XM_006238525.1 | XP_006238587.1 | 7 | 679 | |||
XM_006238526.2 | XP_006238588.1 | 7 | 648 | |||
XM_006238527.2 | XP_006238589.1 | 6 | 581 | |||
XM_006238530.2 | XP_006238592.1 | 5 | 398 | |||
XM_008763898.1 | XP_008762120.1 | 7 | 665 | |||
XM_008763899.1 | XP_008762121.1 | 4 | 337 | |||
XM_008763900.1 | XP_008762122.1 | 6 | 564 | |||
GenBank mRNA | BC162011.1 | - | 5 | 414 | 9091347 |

More Information
Other Species Targeted

Gene Ontology Categories:
