siRNA Details
Gene Symbol: Cdca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115042.2 | NP_001108514.2 | 2 | 217 | ||
XM_008760176.1 | XP_008758398.1 | 2 | 212 | |||
GenBank mRNA | BC097411.1 | - | 2 | 147 | ||
BC158663.1 | - | 2 | 217 | 8369770 | ||
BC158764.1 | - | 2 | 156 | 7111047 |
