siRNA Details
Gene Symbol: Anapc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173983.1 | NP_001167454.1 | 3 | 346 | ||
XM_006243681.2 | XP_006243743.1 | 3 | 645 | |||
XM_006243682.2 | XP_006243744.1 | 3 | 1099 | |||
XM_006243683.2 | XP_006243745.1 | 3 | 619 |
