siRNA Details
Gene Symbol: Gtf3c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109473.1 | NP_001102943.1 | 2 | 769 | ||
XM_006233871.2 | XP_006233933.1 | 2 | 756 | |||
XM_006233872.2 | XP_006233934.1 | 2 | 1546 | |||
XM_008761680.1 | XP_008759902.1 | 3 | 494 |
