siRNA Details
Gene Symbol: Cpeb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245978.2 | XP_006246040.1 | 1 | 929 | ||
XM_006245979.2 | XP_006246041.1 | 1 | 929 | |||
XM_006245980.2 | XP_006246042.1 | 1 | 929 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AK154289, AK162101, AK173229, AK089951.
Gene Ontology Categories:
