siRNA Details
Gene Symbol: Spdef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109530.1 | NP_001103000.1 | 7 | 1632 | ||
XM_006256229.2 | XP_006256291.1 | 6 | 2445 | |||
XM_008772827.1 | XP_008771049.1 | 6 | 1555 | |||
GenBank mRNA | BC166744.1 | - | 6 | 1689 | 7938752 |
