siRNA Details
Gene Symbol: Pdzd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106362.2 | NP_001099832.2 | 4 | 741 | ||
XM_006231449.2 | XP_006231511.1 | 5 | 1347 | |||
XM_008760406.1 | XP_008758628.1 | 5 | 1347 | |||
XM_008760407.1 | XP_008758629.1 | 5 | 1347 |
