siRNA Details
Gene Symbol: LOC691813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759023.1 | XP_008757245.1 | 1843 |
Gene Symbol: Pla2g4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145982.1 | NP_001139454.1 | 13 | 1549 |
