siRNA Details
Gene Symbol: NEWGENE_1594649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759095.1 | XP_008757317.1 | 2 | 189 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NEWGENE_1594649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759095.1 | XP_008757317.1 | 2 | 189 |