siRNA Details
Gene Symbol: ECD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135752.1 | NP_001129224.1 | 15 | 2069 | ||
NM_001135753.1 | NP_001129225.1 | 13 | 1841 | |||
NM_007265.2 | NP_009196.1 | 14 | 1970 | |||
NR_024203.1 | 12 | 1559 | ||||
GenBank mRNA | AK022616.1 | - | 14 | 1918 | ||
AK225519.1 | - | 13 | 1817 | |||
AK315711.1 | - | 15 | 2042 | |||
BC000721.2 | - | 14 | 1934 | 3504956 | ||
CR457407.1 | - | 13 | 1727 | |||
D88208.1 | - | 14 | 1934 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
